This item is now discontinued. Please browse similar items
Thermo Scientific VersaCool 230000005 Autorefill Kit Option
Representative image only.
Thermo Scientific VersaCool Mfr # 230000005 – Item # RK-12128-88
Optimize performance of your Thermo Scientific VersaCool baths
  • Keep your bath running properly with these accessories
  • Keep your high temperature applications running with minimal supervision with the Autofill option
  • 1 year warranty
SPECS & DESCRIPTION
Specifications & Description
  • DescriptionAutorefill Kit Option
  • Warranty1 year
1 Year Warranty
Key Features
  • Keep your bath running properly with these accessories
  • Keep your high temperature applications running with minimal supervision with the Autofill option
  • 1 year warranty
More About this Item
Choose from a wide range of replacement parts and accessories to ensure top performance of your VersaCool baths.
Includes a solenoid valve, both metric and imperial inlet connections, and a control valve to regulate flow into the VersaCool bath. This is key for applications that are running at high temperatures and evaporation in the tank is a concern.