This item is now discontinued. Please browse similar items
Thermo Scientific VersaCool 230000004 Trolley, 17.8" wide x 23.125" long
Representative image only.
Thermo Scientific VersaCool Mfr # 230000004 – Item # RK-12128-87
Optimize performance of your Thermo Scientific VersaCool baths
  • Keep your bath running properly with these accessories
  • VersaCool Trolley adds to the portability of the VersaCool bath
  • 1 year warranty
SPECS & DESCRIPTION
Specifications & Description
  • Wetted MaterialsPainted cold rolled steel
  • DescriptionTrolley, 17.8\" wide x 23.125\" long
  • Warranty1 year
1 Year Warranty
Key Features
  • Keep your bath running properly with these accessories
  • VersaCool Trolley adds to the portability of the VersaCool bath
  • 1 year warranty
More About this Item
Choose from a wide range of replacement parts and accessories to ensure top performance of your VersaCool baths.
Features durable cold-rolled steel platform and four casters— the VersaCool bath is ready for mobile applications. Trolley is set low to the ground, 17.8" wide by 23.125" long and adds about 5/8” in height to the bath if measured from the ground. VersaCool bath mounts directly to the trolley, hardware included, and all four casters can be locked for added safety and stability.