This item is now discontinued. Please browse similar items
Thermo Scientific VersaCool 230000003 Stainless Steel Gable Cover
Representative image only.
Thermo Scientific VersaCool Mfr # 230000003 – Item # RK-12128-86
Optimize performance of your Thermo Scientific VersaCool baths
  • Keep your bath running properly with these accessories
  • Gable cover adds height to the working depth of the VersaCool bath — accommodates two 500-mL Erylenmeyer flasks set side-by-side
  • 1 year warranty
SPECS & DESCRIPTION
Specifications & Description
  • Wetted MaterialsSS
  • DescriptionStainless Steel Gable Cover
  • Warranty1 year
1 Year Warranty
Key Features
  • Keep your bath running properly with these accessories
  • Gable cover adds height to the working depth of the VersaCool bath — accommodates two 500-mL Erylenmeyer flasks set side-by-side
  • 1 year warranty
More About this Item
Choose from a wide range of replacement parts and accessories to ensure top performance of your VersaCool baths.
Designed to add height for taller containers, but still use innovative dripless technology to redirect condensate back into the tank. Stainless steel construction is rugged and resists corrosion. The gable cover should be used in conjunction with the VersaRack to act as a platform for self-standing vessels to assure proper fluid circulation and temperature uniformity.