This item is now discontinued. Please browse similar items
Thermo Scientific Quantech 331775 Borosilicate Cuvettes for Quantech Fluorometer, Pack of Twelve 83075-46
Representative image only.
Thermo Scientific Quantech Mfr # 331775-000 – Item # RK-83075-46
Compatible with your Thermo Scientific Quantech™ Fluorometers
  • 1 year warranty
SPECS & DESCRIPTION
Specifications & Description
  • DescriptionBorosilicate Cuvettes for Quantech Fluorometer, Pack of Twelve 83075-46
  • Warranty1 year
1 Year Warranty
Key Features
  • 1 year warranty
More About this Item
This cuvette is compatible with the Thermo Scientific Quantech™ Fluorometers, modesl 83075-00, -05, -20, and -25.
Cuvettes can be used at two preset heights corresponding to the internationally accepted Z heights so less sample is required. Model 83075-46 borosilicate round cuvette has a 5-mL capacity, and is 12 mm long by 75 mm in diameter.