This item is now discontinued. Please browse similar items
Thermo Scientific Nunc 2650-0020 Cylindrical PP Closed-Dome Tank, 20 gal.
Representative image only.
Thermo Scientific Nunc Mfr # 2650-0020 – Item # RK-50301-53
  • 1 year warranty
SPECS & DESCRIPTION
Specifications & Description
  • MaterialPolypropylene
  • Wall thickness (in)0.25
  • Volume (Gal)20
  • Volume (Liters)76
  • Height (in)32
  • Height (cm)81.28
  • Outside Diameter (OD) (in)16 1/2
  • DescriptionCylindrical PP Closed-Dome Tank, 20 gal.
  • Warranty1 year
1 Year Warranty
Key Features
  • 1 year warranty
More About this Item
The Polypropylene Closed-Dome Tank has a 30-gallon capacity. Mount mixers off-center—a baffle is not required for optimum mixing. Tanks are constructed of polypropylene. You must order sanitary mixer mount 50301-70 with the tank. Cover is included.
Thermo Scientific Nalgene—Reg TM Nalge Thermo Scientific Nunc International