Thermo Scientific Nunc MicroWell™ Microplates, 96-Well, Polypropylene, Non-Treated, Non-Sterile, Conical, 0.45 mL; 120/Cs

Representative image only.
Thermo Scientific Nunc Mfr # 249944 – Item # RK-01929-34
Low binding polypropylene is ideal for homogeneous assays and storage
  • Shared wall design increases the well volume
  • Nontreated polypropylene plates are solvent resistant and autoclavable

PRODUCT OPTIONS

$794.16CAD / Case of 120
Often Bought With…

SPECS & DESCRIPTION

Specifications & Description

  • Number of Wells96
  • Well Volume (mL)0.45
  • Max Working Well Volume (mL)0.4
  • Well ShapeConical
  • Surface TreatmentNon-Treated
  • Lid (Included)No
  • SterileNo
  • Plate ColorNatural
  • Min Temperature (° C)-80
  • Max Temperature (° C)121
  • MaterialPolypropylene
  • Width (mm)86
  • Length (mm)128
  • DescriptionMicroplates, 96-Well, Polypropylene, Non-Treated, Non-Sterile, Conical, 0.45 mL; 120/Cs

Key Features

  • Shared wall design increases the well volume
  • Nontreated polypropylene plates are solvent resistant and autoclavable

More About this Item

Achieve complete sample recovery with the Thermo Scientific™ Nunc™ MicroWell™ 96-well microplates. Plates are ideal for storage as polypropylene has a lower binding capacity so proteins or DNA will not bind. Withstands temperatures from −80° to 121°C (−112° to 250°F).

Choose from conical bottom wells for optimal sample recovery or round bottom wells which are wider for improved mixing.

Warning: This product is not approved or intended for, and should not be used for medical, clinical, surgical or other patient oriented applications.

ACCESSORIES

REVIEWS

Q&A

CUSTOMER SUPPORT

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At800-363-5900
Live chat