Thermo Scientific Nunc Square BioAssay Dishes, Sterile, Treated, 245 x 25 mm; 16/Cs

Representative image only.
Thermo Scientific Nunc Mfr # 240835 – Item # RK-01929-00
Bioassay dish is ideal for agar diffusion assays
  • Constructed of optically clear polystyrene
  • Sterilized by irradiation.
  • External dimensions: 245 x 245 x 25mm

PRODUCT OPTIONS

$434.65CAD / Case of 16

SPECS & DESCRIPTION

Specifications & Description

  • Max Working Well Volume (mL)135
  • Well ShapeSquare
  • Culture Area (cm²)500
  • Surface TreatmentNunclon™ Delta
  • Lid (Included)Yes
  • SterileYes
  • Height (mm)25
  • DescriptionSquare BioAssay Dishes, Sterile, Treated, 245 x 25 mm; 16/Cs

Key Features

  • Constructed of optically clear polystyrene
  • Sterilized by irradiation.
  • External dimensions: 245 x 245 x 25mm

More About this Item

Large, square dishes feature flat bases for uniform distribution of media. Ideal for agar diffusion assays or storage use in screening libraries. Dishes feature stacking ridges and include lids.

REVIEWS

Q&A

CUSTOMER SUPPORT

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At800-363-5900
Live chat