This item is now discontinued. Please browse similar items
Thermo Scientific Nunc Lab-Tek™ Chambered Cover Glass, 1-Well; 16/Pk
Representative image only.
Thermo Scientific Nunc Mfr # 155361 – Item # RK-01838-10
Ideal for in situ analysis—cells can be cultured, fixed, stained, and viewed on the same microscope slide
  • Media chamber securely mounted to No. 1 borosilicate coverglass by non-toxic medical grade silicone adhesive
  • Polystyrene chamber is not removable and requires inverted microscope for viewing
  • Fitted polystyrene cover without top handle minimizes evaporation
SPECS & DESCRIPTION
Specifications & Description
  • Number of Wells1
  • Min Working Well Volume (mL)2.5
  • Max Working Well Volume (mL)4.5
  • Surface TreatmentUntreated
  • MaterialGlass
  • DescriptionChambered Cover Glass, 1-Well; 16/Pk
Sterile
Key Features
  • Media chamber securely mounted to No. 1 borosilicate coverglass by non-toxic medical grade silicone adhesive
  • Polystyrene chamber is not removable and requires inverted microscope for viewing
  • Fitted polystyrene cover without top handle minimizes evaporation
More About this Item
Easily perform microscopic analysis of cell cultures using these miniature chamber systems that have removable media chambers. This eliminates the need for cell transfers from a vessel to a slide. The systems consist of a lid for aseptic manipulation, removable polystyrene (PS) media chamber, and a standard 25 x 75 mm slide that functions as the growth substrate.
Choose from either cell-culture treated Permanox® plastic or glass (soda lime) slides. Both are ideal for analysis of fluorescent stained cell cultures as they do not interfere with fluorescent labels. All Chamber Slide systems are presterilized.
Systems consist of a miniature polystyrene media chamber with a #1 borosilicate cover glass as the growth surface. Chambered cover glasses are sealed to the media chamber using a nontoxic silicone gasket. Ultra-thin (0.13 to 0.17 mm) cover glasses permit viewing of cells at higher magnifications using an inverted microscope. Special German-origin borosilicate glass is ideal for growing fastidious cells such as neuronal cells. Media chambers are not removable.