Cole-Parmer Essentials Narrow-Mouth Plastic Wash Bottle, FEP, 1000mL

Representative image only.
Cole-Parmer Essentials Item # RK-06052-80
Won't contaminate reagents
  • Resistant to most chemicals
  • One-piece closure with spout prevents leaks
  • Autoclavable
  • 6 month warranty

PRODUCT OPTIONS

$459.70CAD / Each

BUY MORE & SAVE

  • 1-4$459.70CAD / Each
  • 5-9$413.73CAD / Each
  • 10+$367.76CAD / Each
Often bought with…

SPECS & DESCRIPTION

Specifications & Description

  • Capacity (mL)1000
  • Capacity (L)1
  • Capacity (oz)33.8
  • Capacity (Gal)0.264
  • MaterialFluorinated Ethylene Propylene (FEP)
  • Cap MaterialFEP
  • Cap / Stopper SizeGL32
  • Cap ColorNatural
  • Bottle ColorNatural
  • Withstands High Temperature (° F)401
  • Withstands Low Temperature (° F)-328
  • AutoclavableYes
  • Neck Inside Diameter (ID) (mm)25
  • Neck Inside Diameter (ID) (in)63/64
  • Outside Diameter (OD) (in)3 59/64
  • Outside Diameter (OD) (cm)10
  • Overall Height (in)8 5/64
  • Overall Height (cm)20.5
  • Overall Depth (in)7 7/8
  • Overall Depth (cm)20
  • Overall Width (in)3 59/64
  • Overall Width (cm)10
  • DescriptionNarrow-Mouth Plastic Wash bottle, 1000ml
  • Warranty6 month
6 Month Warranty

Key Features

  • Resistant to most chemicals
  • One-piece closure with spout prevents leaks
  • Autoclavable
  • 6 month warranty

More About this Item

FEP bottles resist most chemicals and will not contaminate reagents—use to dispense strong solvents. A one-piece closure with spout prevents leaks.

Certificate of Analysis
For a sample Certificate of Analysis, please go to link below under RESOURCES.
If a Lot specific Certificate of Analysis is required, please call 1-800-323-4340 for assistance.

REVIEWS

Q&A

CUSTOMER SUPPORT

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At800-363-5900
Live chat