Thermo Scientific Nalgene 6422-0010 Replacement, polypropylene spigot
Representative image only.
Thermo Scientific Nalgene Mfr # 6422-0010 – Item # RK-06070-22
  • 1 year warranty
PRODUCT OPTIONS
$128.18CAD / Each
BUY MORE & SAVE
  • 1-5$128.18CAD / Each
  • 6-29$115.36CAD / Each
  • 30-119$108.95CAD / Each
  • 120+$102.54CAD / Each
SPECS & DESCRIPTION
Specifications & Description
  • MaterialPolypropylene(PP)
  • Cap MaterialPolypropylene (PP)
  • AutoclavableYes
  • DescriptionReplacement, polypropylene spigot
  • Warranty1 year
1 Year Warranty
Key Features
  • 1 year warranty
More About this Item
Use these spigots with NALGENE carboys. Spigots have 1-1/8"-12 UNF threads and accept 5/8" tubing. Spigot 06073-22 is made of polypropylene and includes an adapter for 1/4" or 1/2" ID tubing. Spigot 06073-23 is made of TEFZEL ETFE; does not include a tubing adapter. NALGENE.