Thermo Scientific Nalgene 2405-0500 PPCO Wash Bottle, 500 mL
Representative image only.
Thermo Scientific Nalgene Mfr # 2405-0500 – Item # RK-06054-10
No leaking while dispensing
  • One-piece closure with spout prevents leaks
  • Autoclavable
  • 1 year warranty
PRODUCT OPTIONS
$163.55CAD / Pkg of 6
BUY MORE & SAVE
  • 1-4$163.55CAD / Pkg of 6
  • 5-9$147.20CAD / Pkg of 6
  • 10+$130.84CAD / Pkg of 6
SPECS & DESCRIPTION
Specifications & Description
  • Capacity (mL)500
  • Capacity (oz)16
  • MaterialPolypropylene(PP)
  • Cap MaterialPolypropylene (PP)
  • Cap / Stopper Size28 mm
  • Cap ColorBlue
  • Bottle ColorTranslucent
  • AutoclavableYes
  • Label TypeBlue Print
  • Withstands High Temperature (° F)250
  • Withstands Low Temperature (° F)-40
  • DescriptionPPCO Wash Bottle, 500 mL
  • Warranty1 year
1 Year Warranty
Key Features
  • One-piece closure with spout prevents leaks
  • Autoclavable
  • 1 year warranty
More About this Item
Polypropylene copolymer (PPCO) bottles are printed with the word “Autoclavable” in bright blue for easy identification. Withstands temperature from -40 to 250°F (-40 to 121°C).