Thermo Scientific Hypercarb HPLC Column, Porous Graphitic Carbon, 3 µm, 100 mm L x 2.1 mm ID

Representative image only.
Thermo Scientific Hypercarb Mfr # 35003-102130 – Item # RK-01891-30
100% porous graphitic carbon provides better separation of structurally similar substances
  • Exceptional retention of very polar analytes
  • Excellent for high temperature applications
  • 100% stainless-steel column hardware
  • pH stable from 0 to 14
  • 1 month warranty

$3,164.84CAD / Each

SPECS & DESCRIPTION

Specifications & Description

  • PhasePorous graphitic carbon
  • Column Length (mm)100
  • Column Inside Diameter (ID) (mm)2.1
  • Column Particle Size (µm)3
  • DescriptionHPLC Column, Porous Graphitic Carbon, 3 µm, 100 mm L x 2.1 mm ID
  • Warranty1 month
1 Month Warranty

Key Features

  • Exceptional retention of very polar analytes
  • Excellent for high temperature applications
  • 100% stainless-steel column hardware
  • pH stable from 0 to 14
  • 1 month warranty

More About this Item

Thermo Scientific™ Hypercarb™ Porous Graphitic Carbon columns have been designed to allow for total pH stability, as well as retention and separation of highly polar components. These columns may be used in both normal and reversed phase HPLC and LC/MS applications.

The unique surface area of Hypercarb columns allow for retention of very polar solutes without the need for ion pair reagents or complex mobile-phase conditions. In addition, these columns are pH stable from 0 to 14, unlike typical silica-based columns. This allows you more choice in buffer selection, while handling high temperatures and pressures. Hypercarb columns also allow for resolution of structurally similar analytes, such as isomers and homologous series, which are not visible in traditional silica-based columns.

REVIEWS

Q&A

CUSTOMER SUPPORT

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At800-363-5900
Live chat