This item is now discontinued. Please browse similar items
Thermo Scientific GENESYS™ Sipper Sample Comparment Accessory for GENESYS™ 100 Series and BioMate 160
Representative image only.
Thermo Scientific Mfr # 840-307200 – Item # RK-02654-46
Enhance the versitility of your spectrophotometer
  • Expand your spectrophotometer capacity and capabilities
  • Sipper sample compartment for sipper pump 02654-30
  • 2 year warranty
SPECS & DESCRIPTION
Specifications & Description
  • Compatible WithGENESYS 140, 150, 180, and 160 BioMate spectrophotometers
  • DescriptionSipper Sample Comparment Accessory for GENESYS™ 100 Series and BioMate 160
Two year warranty
Key Features
  • Expand your spectrophotometer capacity and capabilities
  • Sipper sample compartment for sipper pump 02654-30
  • 2 year warranty
More About this Item
Choose from a wide range of replacement parts and accessories including cuvette holders, sipper pumps, and more suitable for use with the Thermo Scientific™ GENESYS™ visible and UV-visible spectrophotometers. Spectrophotometers sold separately.
Available for sale and use in the United States only.