This item is now discontinued. Please browse similar items
Thermo Scientific Finnpipette 4651060 Pipette , Fixed at 250 uL; Each
Representative image only.
Thermo Scientific Finnpipette Mfr # 4651060 – Item # RK-25014-62
A new level of sophistication created to meet the demands of modern laboratories
  • The first pipette to have an antimicrobial surface treatment to prevent contamination
  • Adjustable finger rest for a customized feel based on your preferences
  • Large, easy-to-read digital display on adjustable-volume models
  • Comfortable, lightweight design with smooth pipetting action
  • Soft-tip ejection
  • Color-coded plunger for quick and easy identification
  • 5-year warranty with web registration
  • 5 year warranty
SPECS & DESCRIPTION
Specifications & Description
  • SterileNo
  • Max Volume (µL)250
  • Max Volume (mL)0.25
  • Max Dispensing (mL)0.25
  • Accuracy (%)±0.40%
  • Reproducibility±0.30%
  • Number Of Channels1
  • DescriptionPipette , Fixed at 250 uL; Each
  • Warranty5 years
5 Year Warranty
Key Features
  • The first pipette to have an antimicrobial surface treatment to prevent contamination
  • Adjustable finger rest for a customized feel based on your preferences
  • Large, easy-to-read digital display on adjustable-volume models
  • Comfortable, lightweight design with smooth pipetting action
  • Soft-tip ejection
  • Color-coded plunger for quick and easy identification
  • 5-year warranty with web registration
  • 5 year warranty