This item is now discontinued. Please browse similar items

Thermo Scientific BioBasic AX HPLC Column, Polyethyleneimine, 5 µm, 50 mm L x 2.1 mm ID

Representative image only.
Thermo Scientific BioBasic Mfr # 73105-052130 – Item # RK-01901-24
Stable polymeric coating is suitable for a wide range of analyte separations
  • Weak anion exchange for multiple charged analytes
  • Can be used for hydrophilic interaction chromatography (HILIC)
  • Proprietary, protein-friendly, ion exchange ligands of polyethyleneimine (PEI) bound to pure deactivated silica
  • Low hydrophobicity
  • 1 month warranty

SPECS & DESCRIPTION

Specifications & Description

  • PhasePolyethyleneimine (PEI)
  • Column Length (mm)50
  • Column Inside Diameter (ID) (mm)2.1
  • Column Particle Size (µm)5
  • Tubing Inside Diameter (ID) (mm)2.1
  • DescriptionAX HPLC Column, Polyethyleneimine, 5 µm, 50 mm L x 2.1 mm ID
  • Warranty1 month
1 Month Warranty

Key Features

  • Weak anion exchange for multiple charged analytes
  • Can be used for hydrophilic interaction chromatography (HILIC)
  • Proprietary, protein-friendly, ion exchange ligands of polyethyleneimine (PEI) bound to pure deactivated silica
  • Low hydrophobicity
  • 1 month warranty

More About this Item

Thermo Scientific™ BioBasic™ HPLC columns provide improved performance for biological samples including peptides, proteins, and other biomolecules. The high purity silica and tailored pore sizes provide superior performance in terms of improved resolution, efficiency, reproducibility, and column life.

BioBasic AX Columns utilize a polymeric secondary-tertiary amine (polyethyleneimine) ligand that is bound to a highly pure, base-deactivated silica that provides separation of proteins, peptides, other anionic species, and polar molecules. The polyethyleneimine bonded phase allows for additional chromatographic options, including hydrophilic interaction chromatography (HILIC) — ideal for separation of sugars and other highly polar species. BioBasic AX columns are i

REVIEWS

Q&A

CUSTOMER SUPPORT

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At800-363-5900
Live chat