This item is now discontinued. Please browse similar items
Thermo Scientific BioBasic 72905-254630 CN Column - Length 250mm, 4.6mm ID, 5µm particle size
Representative image only.
Thermo Scientific BioBasic Mfr # 72905-254630 – Item # RK-01899-80
Provide outstanding reproducibility and efficiency
  • Lower carbon loading for retention of larger proteins and peptides
  • 1 month warranty
SPECS & DESCRIPTION
Specifications & Description
  • PhaseCyano
  • Column Particle Size (µm)5
  • Tubing Inside Diameter (ID) (mm)4.6
  • DescriptionCN Column - Length 250mm, 4.6mm ID, 5µm particle size
  • Warranty1 month
1 Month Warranty
Key Features
  • Lower carbon loading for retention of larger proteins and peptides
  • 1 month warranty
More About this Item
Thermo Scientific BioBasic HPLC columns provide improved performance for biological samples including peptides, proteins, and other biomolecules. The high purity silica and tailored pore sizes provide superior performance in terms of improved resolution, efficiency, reproducibility, and column life.
BioBasic CN Columns contain 300Å pore size silica for maximum performance when used with biomolecules. These columns are ideally suited for the separation of high-hydrophobicity peptides nominally greater than 5,000 Da. BioBasic CN columns provide alternate selectivity and changes in elution order due to interactions through hydrogen bonding, dipole-dipole interactions, and dispersive interactions due to alkyl chain chemistries. The highly pure, deactivated sil