This item is now discontinued. Please browse similar items

Thermo Scientific BioBasic 4 HPLC Column, C4, 5 µm, 150 mm L x 4.6 mm ID

Representative image only.
Thermo Scientific BioBasic Mfr # 72305-154630 – Item # RK-01898-90
Provide excellent analyses of larger proteins and peptides
  • Lower carbon loading for retention of larger proteins and peptides
  • Based on 300Å silica for outstanding biomolecule performance
  • Outstanding reproducibility, efficiency and column lifetime
  • Excellent for LC/MS separations
  • 1 month warranty

SPECS & DESCRIPTION

Specifications & Description

  • PhaseC4
  • Column Length (mm)150
  • Column Inside Diameter (ID) (mm)4.6
  • Column Particle Size (µm)5
  • Tubing Inside Diameter (ID) (mm)4.6
  • Description4 HPLC Column, C4, 5 µm, 150 mm L x 4.6 mm ID
  • Warranty1 month
1 Month Warranty

Key Features

  • Lower carbon loading for retention of larger proteins and peptides
  • Based on 300Å silica for outstanding biomolecule performance
  • Outstanding reproducibility, efficiency and column lifetime
  • Excellent for LC/MS separations
  • 1 month warranty

More About this Item

Thermo Scientific™ BioBasic™ HPLC columns provide improved performance for biological samples including peptides, proteins, and other biomolecules. The high purity silica and tailored pore sizes provide superior performance in terms of improved resolution, efficiency, reproducibility, and column life.

BioBasic 4 Columns contain 300Å pore size silica with lower carbon loading, ideally suited for high-hydrophobicity peptides nominally greater than 5,000 Da. Provide less retention of hydrophobic peptides due to the shorter alkyl length.

REVIEWS

Q&A

CUSTOMER SUPPORT

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At800-363-5900
Live chat