This item is now discontinued. Please browse similar items
Thermo Scientific Barnstead D50253 Ultrapure deionization cartridge, 680 grains as NaCl
Representative image only.
Thermo Scientific Barnstead Mfr # D50253 – Item # RK-99275-62
  • 1 year warranty
SPECS & DESCRIPTION
Specifications & Description
  • Capacity (grains)680 as Sodium Chloride
  • Compatible withNanopure Infinity
  • FunctionUltrapure, for high resistivity
  • DescriptionUltrapure deionization cartridge, 680 grains as NaCl
  • Warranty1 year
1 Year Warranty
Key Features
  • 1 year warranty
More About this Item
Replace cartridges regularly for best quality and performance.