Thermo Scientific Barnstead D5021 posttreatment organic-free cartridges
Representative image only.
Thermo Scientific Barnstead Mfr # D5021 – Item # RK-01506-05
  • 1 year warranty
$419.92CAD / Each
SPECS & DESCRIPTION
Specifications & Description
  • Capacity (grains)370 as Sodium Chloride
  • Compatible withQuad-style NANOpure™, E-pure™ systems
  • FunctionRemoves organics to <20 ppb organic carbon; use for HPLC, BOD, pH, and cell cultures
  • Descriptionposttreatment organic-free cartridges
  • Warranty1 year
1 Year Warranty
Key Features
  • 1 year warranty
More About this Item
Post-treat to polish your water—make it organic free.