Thermo Scientific Barnstead D400499 Water Purification Cartridge, High-Capacity
Representative image only.
Thermo Scientific Barnstead Mfr # D400499 – Item # RK-99292-91
  • 1 year warranty
PRODUCT OPTIONS
$1,517.59CAD / Each
SPECS & DESCRIPTION
Specifications & Description
  • DescriptionWater Purification Cartridge, High-Capacity
  • Warranty1 year
1 Year Warranty
Key Features
  • 1 year warranty
More About this Item
Clean your still less often by reducing the level of scaling ions entering your still. Replacement cartridges are compatible with both single- and dual-cartridge deionizers.
Capacity of up to 2400 grains as CaCO3.