Thermo Scientific Barnstead Smart2Pure 6 UV Water Purification System, 6L/Hr

Representative image only.
Thermo Scientific Barnstead Mfr # 50129885 – Item # RK-99259-12
Both Type I and Type II water in a complete space-saving design
  • Display can be tilted for optimal reading
  • Variable-flow control for dispensing of Type I water
  • Quick-change cartridge design simplifies self maintenance, even in mid-operations
  • 1 year warranty

PRODUCT OPTIONS

$15,477.96CAD / Each

SPECS & DESCRIPTION

Specifications & Description

  • ASTM Water GradeType I and Type II
  • Length (\")12
  • Capacity6 L/hr
  • Width (InchShort)15 3/4
  • Height (in)21 29/64;21.46
  • Max Production (L/hour)6
  • Max Flow Rate (L/min)0.6
  • Max Flow Rate (L/hour)36
  • Max Flow Rate (GPH)9.5
  • Inlet Port3/4\" NPT
  • With UV LampYes
  • With UltrafilterNo
  • Resistivity Threshold18.2 MΏ-cm
  • TOC Monitor1 to 5ppb
  • Conductivity Threshold0.055 µS/cm
  • Sourcetap
  • TOC Threshold5ppb
  • Feed Water Requirements (Source)Potable Water according to DIN 2000
  • Type I Water (Resistivity)18.2 MΩ-cm
  • Type I Water (TOC)1 to 5 ppb
  • Feed Water Requirements (TOC)1 to 5 ppb
  • Type I Water (Conductivity)0.055 µS/cm
  • Feed Water Requirements (Bacteria)<0.01 CFU/mL
  • Type I Water (Bacteria)<0.01 CFU/mL
  • Type I Water (Endotoxins)0.005EU/mL
  • Feed Water Requirements (pressure)14.5 to 87 psi (1 to 6 bar)
  • Type II Water (Resistivity)10 to 15 MΩ-cm
  • Feed Water Requirements (Temperature)2-35⁰C
  • Type II Water (TOC)1 to 5 ppb
  • Type II Water (Conductivity)0.067 to 0.1 µS-cm
  • Min Inlet Water Temperature (° F)35.6
  • Min Inlet Water Temperature (° C)2
  • Min Inlet Pressure (PSI)14.5
  • Max Inlet Pressure (PSI)87
  • Min Inlet Pressure (bar)1
  • Max Inlet Pressure (bar)6
  • Min Operating Pressure (PSI)14.5
  • Max Operating Pressure (PSI)87
  • Min Operating Pressure (bar)1
  • Max Operating Pressure (bar)6
  • Pressure (bar)6
  • Pressure (PSI)87
  • Depth (in)15.75
  • Width (in)15.75
  • Height (cm)54.5
  • Depth (cm)40
  • Width (cm)40
  • DescriptionSmart2Pure, 6 L/hr, UV
  • Warranty1 year
1 Year WarrantyConformite EuropeenneCanadian Standards Association

Key Features

  • Display can be tilted for optimal reading
  • Variable-flow control for dispensing of Type I water
  • Quick-change cartridge design simplifies self maintenance, even in mid-operations
  • 1 year warranty

More About this Item

Barnstead Smart2Pure systems deliver up to 72 L/day of Type I ultrapure water or Type II pure water from a tap feed water supply. Depending on your requirements, choose from three production capacities—3, 6, or 12 L/hr. The 12 L/hr systems let you safely store high purity water in either a 30- or 60-L reservoir, easily accessible when you need it!

These compact systems are fully customizable to support your tissue culture, PCR, electrophoresis, atomic absorption, ICP, ICP/MS, and chromatography needs. In addition, applications requiring Type II water such as reagent preparation and glassware rinsing can be met. The RO membrane with pretreatment cartridge is separate from the DI cartridge, ensuring you only replace each cartridge as needed.

The systems with UV option offer UV-oxidation (185/254 nm) for reducing the content of micoorganisms and their metabolites.

An internal 6-L tank with vent filter is included.

Pretreatment filter is required (sold seperately)

Warning: This product is not approved or intended for, and should not be used for medical, clinical, surgical or other patient oriented applications.

ACCESSORIES

REVIEWS

Q&A

CUSTOMER SUPPORT

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At800-363-5900
Live chat