This item is now discontinued. Please browse similar items
Thermo Scientific Aquabath 19000-17Q Stainless Steel Cover 20 and 28L Aquabaths
Representative image only.
Thermo Scientific Aquabath Mfr # 19000-17Q – Item # RK-03710-86
Reduce heat loss
  • Constructed of stainless steel
  • 1 year warranty
SPECS & DESCRIPTION
Specifications & Description
  • Reservoir Capacity (L)20
  • DescriptionStainless Steel Cover 20 and 28L Aquabaths
  • Warranty1 year
1 Year Warranty
Key Features
  • Constructed of stainless steel
  • 1 year warranty
More About this Item
These water bath covers are compatible with Lab-Line AquaBath Brand water baths. Covers ensures temperatures are maintained while in use. Water baths are sold separately.