This item is now discontinued. Please browse similar items
Thermo Scientific Aquabath 18000AQ 5L Analog Utility Bath - 120V
Representative image only.
Thermo Scientific Aquabath Mfr # 18000AQ – Item # RK-03710-09
Precise temperature control to 100°C
  • Built-in safety - dual thermostats prevent overheating
  • Simple maintenance - rounded stainless steel interiors & drain pumps
  • 2 year warranty
SPECS & DESCRIPTION
Specifications & Description
  • Bath TypeAnalog Water Bath
  • Max Temperature (° C)100
  • Min Temperature (° C)Ambient
  • Power (VAC)115
  • Description5L Analog Utility Bath - 120V
  • Warranty2 year
Two year warrantyConformite EuropeenneCanadian Standards Association
Key Features
  • Built-in safety - dual thermostats prevent overheating
  • Simple maintenance - rounded stainless steel interiors & drain pumps
  • 2 year warranty
More About this Item
Analog baths offer easy-to-use analog control and hydraulic thermostat ideal for fixed set-point applications. Digital models feature PID contol and digital temperature set and readout selectable display, resolution of 0.1°C. All models feature stainless-steel exterior shell, reservoir, and sample tray. Hinged cover lifts to 90-degree stay-open position or can be removed completely.