This item is now discontinued. View the direct replacement: RK-00653-09 or browse similar items
Thermo Scientific 910760 pH Buffer Solution, 7.00, Yellow; 5 x 60 mL Bottles
Representative image only.
Thermo Scientific Mfr # 910760 – Item # RK-55350-44
Common calibration values are olor-coded for ease of use
  • Choose from pint-size or portable 60-mL bottles
  • Traceable to NIST standards
SPECS & DESCRIPTION
Specifications & Description
  • Buffer Value7
  • Buffer Kit Values7
  • Volume (mL)60
  • ColorYellow
  • DescriptionpH Buffer Solution, 7.00, Yellow; 5 x 60 mL Bottles
Key Features
  • Choose from pint-size or portable 60-mL bottles
  • Traceable to NIST standards
More About this Item
Thermo Scientific Orion pH buffers are available in 475-mL (pint) bottles and in portable 5 x 60 mL convenience packs. Portable pH buffers are easier to carry than larger bottles, minimize wasted buffer, and provide more volume than pouches.
Each buffer is color-coded for easy identification and packaged with buffer/temperature tables for accurate calibrations. Buffers are standardized against NIST-traceable pH references for superior accuracy.