This item is now discontinued. Please browse similar items

Thermo Scientific 05EREETSA Explosion Proof Undercounter Refrigerator, 5.5 cu ft; 115V

Representative image only.
Thermo Scientific Mfr # 05EREETSA – Item # RK-44200-20
Reduce the risk of explosion with a spark-free interior and exterior
  • Designed for use in Class I, Groups C & D & Class I, Zone 1, Group IIB hazardous environments
  • CFC-free insulation and refrigerant
  • 15 month warranty

SPECS & DESCRIPTION

Specifications & Description

  • Refrigerator Capacity (cu ft)5.5
  • Refrigerator Capacity (Liters)155.7
  • Freezer Capacity (cu ft)17.2
  • Freezer Capacity (Liters)581
  • Min Temperature - Refrigerator (° F)33
  • Max Temperature - Refrigerator (° F)53
  • Min Temperature - Refrigerator (° C)1
  • Max Temperature - Refrigerator (° C)12
  • Min Temperature - Freezer (° F)-40
  • Max Temperature - Freezer (° F)14
  • Min Temperature - Freezer (° C)-40
  • Max Temperature - Freezer (° C)-10
  • Refrigerator Interior Width (in)20
  • Refrigerator Interior Height (in)27 3/4
  • Refrigerator Interior Depth (in)17 1/2
  • Refrigerator Interior Width (cm)50.8
  • Refrigerator Interior Height (cm)70.5
  • Refrigerator Interior Depth (cm)44.5
  • Exterior Width (in)23 5/8
  • Exterior Height (in)33 1/2
  • Exterior Depth (in)23 1/2
  • Exterior Width (cm)60
  • Exterior Height (cm)85.1
  • Exterior Depth (cm)60
  • Number of Doors1
  • Number of Shelves3
  • Cabinet ConstructionEnameled Steel
  • Power (VAC)115
  • Power (Hz)60
  • Power (amps)1.33
  • Power (watts)160
  • Power (Breaker)15 A
  • DescriptionExplosion Proof Undercounter Refrigerator, 5.5 cu ft; 115V
  • Warranty15 month
15 Month WarrantyCanadian Standards Association

Key Features

  • Designed for use in Class I, Groups C & D & Class I, Zone 1, Group IIB hazardous environments
  • CFC-free insulation and refrigerant
  • 15 month warranty

More About this Item

These explosion proof refrigerators are particularly suited for safe storage in hazardous environments. Featuring thermostat and compressor components that are isolated within a vapor-proof enclosure, and electrical connections that are hardwired, these units will prevent sparking and accidental disconnection. All models feature manual defrost.

REVIEWS

Q&A

CUSTOMER SUPPORT

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At800-363-5900
Live chat