This item is now discontinued. Please browse similar items
Testo Smart Probe VAC Kit for use with Smartphone or Tablet
Representative image only.
Testo Mfr # 0563 0003 – Item # RK-30009-47
View HVAC measurement on your Smart device from up to 60 feet away!
  • Four instruments in one convenient kit for checking ventilation systems
  • Use the Smart Probes app to read and create custom reports
  • Save and share measurements as a PDF or Excel® file
  • Perform difficult calculations automatically
  • Multi-color LED displays low battery warning and Bluetooth® connectivity
  • 2 year warranty
SPECS & DESCRIPTION
Specifications & Description
  • Max Air Velocity (ft/min)6000
  • Min Air Velocity (ft/min)0
  • Temperature Resolution0.1°F
  • Min IR Temperature (° F)-22
  • Min IR Temperature (° C)-30
  • Max IR Temperature (° F)482
  • Max IR Temperature (° C)250
  • Max RH (%)100
  • Min RH (%)0
  • RH Resolution0.1%
  • BatteryThree AAA batteries (included)
  • DescriptionWireless VAC Smart Probe Kit
  • Warranty2 year
Two year warrantyApple App StoreGoogle Play App
Key Features
  • Four instruments in one convenient kit for checking ventilation systems
  • Use the Smart Probes app to read and create custom reports
  • Save and share measurements as a PDF or Excel® file
  • Perform difficult calculations automatically
  • Multi-color LED displays low battery warning and Bluetooth® connectivity
  • 2 year warranty
More About this Item
The VAC kit provides all the tools necessary to measure air velocity, temperature, relative humidity, wet bulb/dew point, and calculate volume flow of your multi-outlet ventilation system – ideal for the HVAC professional. Kit includes the Testo 405i Smart Probe hot-wire anemometer, Testo 410i Smart Probe vane anemometer, Testo 605i Smart Probe thermohygrometer, and Testo 805i Smart Probe infrared thermometer. Both anemometers measure air velocity, volume flow and temperature. The thermohygrometer measures relative humidity and temperature as well as automatically calculates dew point and wet bulb temperature. The infrared thermometer provides non-contact temperature readings, ideal for checking breakers, motors, ducts, and registers from a distance. A circular laser pattern ensures accurate target measurement.
Easily monitor and transmit all measured data wirelessly via Bluetooth® to your smartphone or tablet from up to 60 feet away. With the free Testo Smart Probes app, you can display measurements from up to six Smart Probes at the same time, visualize measurement value changes as a graph or a table, and perform automatic calculations. Easily turn your data into a custom report, attach images created with the app, and send via email as a PDF or Excel® file.
Available for sale in the United States and Canada only.