This item is now discontinued. Please browse similar items
Testo Smart Probe Hydronic Heating System Kit for use with Smartphone or Tablet
Representative image only.
Testo Mfr # 0563 0004 – Item # RK-30009-45
View HVAC measurement on your Smart device from up to 60 feet away!
  • Three instruments in one convenient kit for checking hydronic heating system
  • Use the Smart Probes App to read and create custom reports
  • Save and share measurements as PDF or Excel® files
  • Perform difficult calculations automatically
  • Multi-color LED displays low battery warning and Bluetooth® connectivity
  • 2 year warranty
SPECS & DESCRIPTION
Specifications & Description
  • Accuracy±0.02\" WC from 0 to 0.4\" WC;
  • Min Pressure (in WC)-60
  • Max Pressure (in WC)60
  • Min Temperature (° F)-40
  • Max Temperature (° F)482
  • Min Temperature (° C)-40
  • Max Temperature (° C)250
  • Unit Of Measurementmbar, hPa, Pa, mmHg, \" Hg, \" WC, psi, mm WC, m/sec, ft/min, m3/hr, cfm, L/sec
  • DisplaySmart Phone App
  • Battery TypeThree AAA batteries
  • Resolution0.01\" WC
  • Power SourceBattery
  • DescriptionSmart & Wireless Hydronic Heating System Smart Probe Kit
  • Warranty2 year
Two year warrantyApple App StoreGoogle Play App
Key Features
  • Three instruments in one convenient kit for checking hydronic heating system
  • Use the Smart Probes App to read and create custom reports
  • Save and share measurements as PDF or Excel® files
  • Perform difficult calculations automatically
  • Multi-color LED displays low battery warning and Bluetooth® connectivity
  • 2 year warranty
More About this Item
Use the heating system kit to perform timed pressure leak-down tests on pipe systems, measure temperature of pipes in heating and cooling systems, and photograph objects with your smartphone or tablet. With a free Smart Phone app all measured data are transmitted to your smart phone/tablet so the data can be read, turned into a report, or sent by an email. Besides giving you the measured values, the Smart Probes app can visualize measurement value changes as a graph or a table as well as perform calculations such as evaporation and condensation temperatures, superheating and subcooling, volume flow measurement in ducts and at outlets. Easily support data reports with images created with the app and sent as PDF or Excel files.
Testo hydronic heating system kit includes three instruments—clamp thermometer, infrared thermometer, and differential pressure manometer—for a convenient heating system check. Use clamp thermometer on 1/4 to 1-1/2" diameter pipes to measure temperature in heating and cooling systems. IR thermometer is ideal for checking breakers, motors, ducts, and registers from a distance. A circular laser pattern ensures accurate target measurement. Differential pressure manometer measures manifold pressure and pressure drops in building or ventilation systems. A measurement menu in the Smart Probes app includes pressure drop alarms. Magnetic back holds instrument in place for hands-free operation.
Available for sale in the United States and Canada only!