Testo 605i Smart Probe Thermohygrometer for use with Smartphone or Tablet
Representative image only.
Testo Mfr # 0560 2605 03 – Item # RK-30009-42
View HVAC measurement on your Smart device from up to 60 feet away!
  • Use the Smart Probes app to read and create custom reports
  • Save and share measurements as a PDF or Excel® file
  • Perform difficult dew point and wet bulb temperature calculations automatically
  • Multi-color LED displays low battery warning and Bluetooth® connectivity
  • 2 year warranty
$228.38CAD / Each
SPECS & DESCRIPTION
Specifications & Description
  • Min RH Range (%)0
  • Max RH Range (%)100
  • RH Accuracy±(1.8% RH + 3% of m.v.) at 77 °F (5 to 80% RH)
  • RH Resolution (%)0.1
  • Min Temperature (° F)-4
  • Min Temperature (° C)-20
  • Max Temperature (° F)140
  • Max Temperature (° C)60
  • Temperature Accuracy±1.4°F (-4 to 32°F), ±0.9°F (32 to 140°F)
  • Temperature Resolution0.1°
  • Battery3 AAA (included)
  • DescriptionSmart Probe Thermohygrometer
  • Warranty2 year
Two year warrantyApple App StoreConformite EuropeenneGoogle Play App
Includes
Three AAA batteries and certificate of calibration.
Key Features
  • Use the Smart Probes app to read and create custom reports
  • Save and share measurements as a PDF or Excel® file
  • Perform difficult dew point and wet bulb temperature calculations automatically
  • Multi-color LED displays low battery warning and Bluetooth® connectivity
  • 2 year warranty
More About this Item
This compact, rugged, easy-to-use wireless thermohygrometer provides accurate measurement of relative humidity, dew point, and temperature in rooms, ducts, registers, and chambers – ideal for the HVAC professional. The small-diameter probe is ideal for in-duct measurements.
Easily monitor and transmit all measured data wirelessly via Bluetooth® to your smartphone or tablet from up to 60 feet away. With the free Testo Smart Probes app, you can display measurements from up to six Smart Probes at the same time, visualize measurement value changes as a graph or a table, and perform automatic calculations. Easily turn your data into a custom report, attach images created with the app, and send via email as a PDF or Excel® file.
Combine this probe with the 405i Smart Probe hot-wire anemometer, 410i Smart Probe vane anemometer, and 805i Smart Probe infrared thermometer to monitor HVAC-R applications.
Available for sale in the United States and Canada only.