This item is now discontinued. Please browse similar items
Testo 0613 3211 Thermistor Frozen Food Probe
Representative image only.
Testo Mfr # 0613 3211 – Item # RK-37803-60
Different probe styles accommodate a variety of applications
  • NTC temperature sensor
  • Can be screwed directly into frozen food, no pre-drilling required
  • EN 13485 and HACCP-compliant
  • Measuring range of -50 to +140°C
  • 2 year warranty
SPECS & DESCRIPTION
Specifications & Description
  • DescriptionThermistor Frozen Food Probe
Two year warranty
Key Features
  • NTC temperature sensor
  • Can be screwed directly into frozen food, no pre-drilling required
  • EN 13485 and HACCP-compliant
  • Measuring range of -50 to +140°C
  • 2 year warranty
More About this Item
Thermistors exhibit a greater senstitivity and accuracy in the biological range. Probes encase a ceramic element that generally decreases in resistance as the temperature increases.
The frozen food probe (NTC) is used primarily for measuring the core temperature of frozen food. Thanks to a special handle and its helical tip, it can be screwed directly into frozen food without the need for pre-drilling. The probe is EN 13485 and HACCP-compliant, making it particularly practical for use in the food sector when connected to a corresponding measuring instrument (e.g. testo 110). The special handle is heat-resistant up to a temperature of +70 °C.