This item is now discontinued. Please browse similar items

Testo 0613 2411 Thermistor Food Probe -58 to 300°F (-50 to 148°C)

Representative image only.
Testo Mfr # 0613 2411 – Item # RK-37803-59
Different probe styles accommodate a variety of applications
  • NTC temperature sensor
  • EN 13485 and HACCP-compliant
  • Special handle with reinforced PUR cable
  • Measuring range of -25 to +150 °C
  • 2 year warranty

SPECS & DESCRIPTION

Specifications & Description

  • DescriptionThermistor Food Probe -58 to 300°F (-50 to 148°C)
Two year warranty

Key Features

  • NTC temperature sensor
  • EN 13485 and HACCP-compliant
  • Special handle with reinforced PUR cable
  • Measuring range of -25 to +150 °C
  • 2 year warranty

More About this Item

Thermistors exhibit a greater senstitivity and accuracy in the biological range. Probes encase a ceramic element that generally decreases in resistance as the temperature increases.

Robust food penetration probe with special handle, IP 65, reinforced cable (PUR) and reinforced protection against buckling

REVIEWS

Q&A

CUSTOMER SUPPORT

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At800-363-5900
Live chat