This item is now discontinued. Please browse similar items
Testo 0560 7207 Compact RTD/Thermistor Thermometer
Representative image only.
Testo Mfr # 0560 7207 – Item # RK-86460-07
Professional single channel thermometer designed for the demands of the laboratory but rugged enough for the industrial use.
  • Accepts both RTD and thermistor probes
  • Select from an impressive range of optional probes
  • 2 year warranty
SPECS & DESCRIPTION
Specifications & Description
  • Meter Measurement TypeMulti-Parameter
  • Number Of Probe Channels1
  • Types Of Probes AcceptedRTD, Thermistor
  • Min Temperature (° F)-150
  • Max Temperature (° F)1472
  • Min Temperature (° C)-100
  • Max Temperature (° C)800
  • AccuracyRTD: ±0.2% of reading; ±0.4°F (0.2°C) above 800°F (426°C) and below 200°F (93°C); Thermistor ±0.4°; ±0.5° between 104° to 180°F (40° to 80°C); ±0.7° between 180° to 260°F (80° to 125°C); ±0.9° above 260°F (125°C) and below -13°F (-25°C)
  • Resolution0.1°
  • Low Battery IndicatorYes
  • Display TypeLCD
  • Height (in)7
  • Width (in)2 1/2
  • Depth (in)1 1/2
  • Battery9v Battery (1)
  • DescriptionCompact RTD/Thermistor Thermometer
  • Warranty2 year
Two year warranty
Key Features
  • Accepts both RTD and thermistor probes
  • Select from an impressive range of optional probes
  • 2 year warranty
More About this Item
The wide measuring range and accurate four-wire technology makes this thermometer an ideal instrument for everyday laboratory work. The meter accepts a wide range of optional probes (meter does not come with a probe) that can measure temperature within seconds. Features user-adjustable audible alarm limits, min/max/hold functions, a large two-line backlit display, and basic status such as battery life.
Optional carrying case 10323-12 protects the instrument from moisture, dirt, and impact — yet allows easy operation and viewing of the display. Readings can be printed on-site with optional wireless printer 37486-12 to provide a permanent record.