This item is now discontinued. Please browse similar items
Testo 0560 1108 Compact Thermistor Thermometer with Wireless Capability
Representative image only.
Testo Mfr # 0560 1108 – Item # RK-37803-54
Highly accurate and versatile-specially designed for taking measurements in rough conditions such as refrigerated rooms.
  • Meter requires the probes to be ordered separately from an impressive range of options
  • 2 year warranty
SPECS & DESCRIPTION
Specifications & Description
  • Meter Measurement TypeThermistor
  • Number Of Probe Channels1
  • Types Of Probes AcceptedThermistor
  • Min Temperature (° F)-58
  • Max Temperature (° F)530
  • Min Temperature (° C)-50
  • Max Temperature (° C)276
  • Accuracy±0.5°F (±0.5°C)
  • Resolution0.1°
  • Height (in)7
  • Width (in)2 1/2
  • Depth (in)1 1/2
  • Battery9v Battery (1)
  • DescriptionCompact Thermistor Thermometer with Wireless Capability
  • Warranty2 year
Two year warranty
Key Features
  • Meter requires the probes to be ordered separately from an impressive range of options
  • 2 year warranty
More About this Item
This meter has an impressive range of optional probes (meter does not come with a probe) that can measure temperature within seconds. Features user-adjustable audible alarm limits, min/max/hold, a large two-line backlit display, and basic status such as battery life. In addition to standard probes with a wire, this meter can be easily upgraded anytime to wireless probes for use up to a distance of 20 meters. A maximum of one wired and one wireless probe can be read and displayed by the meter. To add this feature, order wireless upgrade kit 37486-02 (includes one wireless handle and module) along with the wireless thermistor probe.
Optional carrying case 10323-12 protects the instrument from moisture, dirt, and impact — yet allows easy operation and viewing of the display.