This item is now discontinued. Please browse similar items
TestCoat 312055-QNIX1200 Coating Thickness Gauge, FE; 120 mils
Representative image only.
TestCoat Mfr # 312055-QNIX1200 – Item # RK-59784-11
Reliable results with probe free design
  • Measures non-magnetic coatings on steel and iron
  • Easy and precise with no calibration or probe change required
  • Wide measuring range of 120 mils
  • 3 year warranty
SPECS & DESCRIPTION
Specifications & Description
  • DescriptionAccessory Ferrous Only Quanix 1200 Basic
  • Warranty3 year
3 Year Warranty
Key Features
  • Measures non-magnetic coatings on steel and iron
  • Easy and precise with no calibration or probe change required
  • Wide measuring range of 120 mils
  • 3 year warranty
More About this Item
As patented electronic Paint Thickness Gauge/ Meter with integrated probe for steel and iron, the QuaNix/QNix 1200 with a clear readable duplex LCD offers an unusually extensive spectrum of use and a large measuring range, up to 120 mils. The flat design of this gauge allows for measurements at difficult-to-access points. Simple and flexible use, even at wide stretching surfaces.