Tensioning Tool for 18 to 50 lb Cable Zip Ties, Heavy Duty Aluminum
Representative image only.
Item # RK-06830-00
Ideal for high-volume wire tie applications
  • Bundles wires efficiently while reducing operator fatigue
  • Adjustable knob allows selection of tightness and locks for precise, repeatable tension
  • Rugged, lightweight cast aluminum body
  • Cable zip ties are automatically cut when preset tension level is reached
$581.99CAD / Each
SPECS & DESCRIPTION
Specifications & Description
  • MaterialAluminum
  • DescriptionTensioning Tool for 18 to 50 Pound Cable/Zip Ties, Aluminum
Key Features
  • Bundles wires efficiently while reducing operator fatigue
  • Adjustable knob allows selection of tightness and locks for precise, repeatable tension
  • Rugged, lightweight cast aluminum body
  • Cable zip ties are automatically cut when preset tension level is reached
More About this Item
Comfort and reliability are the benefits of this shock-resistant, sturdy aluminum cable tie gun. It tightens plastic cable zip ties, then automatically cuts them when the preset tension is reached. A non-slip tension knob locks to ensure repeatable tension, tie after tie.
To lessen fatigue and increase comfort for smaller hands, the distance between the trigger and handle is minimized to lower the trigger force necessary for operation. The rugged, lightweight cast aluminum body and reliable internal mechanisms makes this tool ideal for use in production environments. The blade and tool geometry improves cutting capability for smoother, more precise cuts.
Use with miniature (18 lb), intermediate (40 lb), and standard ties (50 lb) ties with a maximum width 0.3” (7.6 mm).