This item is now discontinued. Please browse similar items
Cole-Parmer FHTFHI High Temperature Bath Fluid, 60 to 250 C; 18 L
Representative image only.
Cole-Parmer Item # RK-12122-99
Enhance the performance of your Cole-Parmer® baths and circulators
  • Enhances bath temperature uniformity and stability
  • Safe and non-toxic high temperature bath fluid that is versatile and cost effective
  • 1 year warranty
SPECS & DESCRIPTION
Specifications & Description
  • Compatible WithAll water baths
  • DescriptionHigh Temperature Bath Fluid, 60 to 250 C; 18 L
  • Warranty1 year
1 Year Warranty
Key Features
  • Enhances bath temperature uniformity and stability
  • Safe and non-toxic high temperature bath fluid that is versatile and cost effective
  • 1 year warranty
More About this Item
Lids, trays, temperature probes, and other accessories for your bath.
Recommended temperature range for closed systems is 0 to 315°C, and the recommended range for open systems is 20 to 150°C.
The Flash point of this bath fluid is 180°C (356°F) and the Autoignition temperature is 330°C (626°F).
Like many heat transfer fluids, may ignite if exposed to hot surfaces, sparks, flames.