This item is now discontinued. Please browse similar items
Taylor 509 Connoisseur Series Gourmet Candy / Deep Fry Analog Dial Thermometer with 7" Stem
Representative image only.
Taylor Mfr # 509 – Item # RK-90001-38
Use this thermometer to ensure a consistent final product every time!
  • Adjust immersion depth with the included clip
  • Temperature range 100 to 400F
  • Oversized 3'' face with 2 1/4'' dial
  • Silicone coated pan clip won't scratch pans
  • 7'' stainless steel stem
  • 1 year warranty
SPECS & DESCRIPTION
Specifications & Description
  • Min Temperature (° F)100
  • Max Temperature (° F)400
  • Accuracy2%
  • Stem Length (in)6
  • DescriptionGourmet Candy / Deep Fry Analog Dial Thermometer with 7\" Stem
  • Warranty1 year
1 Year Warranty
Key Features
  • Adjust immersion depth with the included clip
  • Temperature range 100 to 400F
  • Oversized 3'' face with 2 1/4'' dial
  • Silicone coated pan clip won't scratch pans
  • 7'' stainless steel stem
  • 1 year warranty
More About this Item
An essential addition to commercial kitchens, this thermometer allows you to easily check the temperatures of delicate candy mixes or fryer oils so your final food products come out just right.
Connoisseur Candy/Deep Fry Thermometer, 3" head with 2-1/4" viewing dial, 100° to 400°F temperature range, stainless steel construction with silicone grips, stem with silicone coated pan clip