Supco 6" Circular Chart Recorder Paper, 31 Day, 0 to 250°F; 60/Box
Representative image only.
Supco Mfr # CR1000-19 – Item # RK-80051-61
For Supco brand chart recorders
  • Circular and strip charts
  • For CR87HT series recorders
  • 1 year warranty
PRODUCT OPTIONS
$84.13CAD / Box of 60
SPECS & DESCRIPTION
Specifications & Description
  • Chart TypeCircular
  • Chart Speed31 day
  • Chart Diameter (in)6
  • Max Temperature (° F)250
  • Min Temperature (° F)0
  • Max Temperature (° C)121
  • Min Temperature (° C)-18
  • Description6\" Circular Chart Recorder Paper, 31 Day, 0 to 250°F; 60/Box
  • Warranty1 year
1 Year Warranty
Key Features
  • Circular and strip charts
  • For CR87HT series recorders
  • 1 year warranty
More About this Item
Choose the paper compatible with your Supco chart recorder and application needs.