Stuart SHM/TOOL Tool Kit for Stainless Steel Homogenizer Probe Disassembly
Representative image only.
Stuart Item # RK-04720-13
Make the most of your homogenizer
  • Probes available in different sizes and materials
  • Stand for benchtop use
  • Parts and adapters to expand capabilities
  • Use to disassemble stainless probes for change out and cleaning
PRODUCT OPTIONS
Call For Price
SPECS & DESCRIPTION
Specifications & Description
  • Compatible WithStuart SHM/X Series stainless steel probes
  • DescriptionTool Kit for Stainless Steel Homogenizer Probe Disassembly
Key Features
  • Probes available in different sizes and materials
  • Stand for benchtop use
  • Parts and adapters to expand capabilities
  • Use to disassemble stainless probes for change out and cleaning
More About this Item
Stainless steel rotor probes work well with large volumes, and disposable polycarbonate probes reduce cross-contamination (autoclave and reuse up to seven times!). Ideal for tissue homogenisation, cell disruption, emulsions, suspensions, and protein extraction.