This item is now discontinued. View the direct replacement: RK-07387-37 or browse similar items
18.802 Straight: Very Fine Tips; 1-mm smooth tips
Representative image only.
Aven Tools Mfr # 18.802 – Item # RK-07290-04
SPECS & DESCRIPTION
Specifications & Description
  • Overall Length (in)5
  • Overall Length (cm)12.7
  • MaterialCeramic
  • SterileYes
  • DescriptionStraight: Very Fine Tips; 1-mm smooth tips
More About this Item
These tweezers meet the highest requirements for use in semiconductor manufacturing, cleanrooms, chemical laboratories, biotechnology, and circuit board manufacturing. Features an extremely smooth surface, and very low coefficient of friction. Ceramic tips are highly resistant to all strong acids (except hydrofluoric); 100% non-magnetic, non-rusting, and non-static; temperature resistant up to 1500°C.