This item is now discontinued. Please browse similar items
Stoppers, Platinum-Cured Silicone, Standard Size 12
Representative image only.
Item # RK-62995-30
SPECS & DESCRIPTION
Specifications & Description
  • Standard Size Number12
  • Top Outside Diameter (OD) (mm)63.9
  • Bottom Outside Diameter (OD) (mm)54
  • ColorWhite
  • MaterialPlatinum-cured Silicone
  • DescriptionStoppers, Platinum-Cured Silicone, Standard Size 12
More About this Item
Ultra-pure, platinum-cured silicone stoppers are ideal for high-purity applications. They exhibit excellent resistance to most chemicals except strong acid. Inherent resistance to cracking and spallation significantly reduces any possibility of particulate contamination. Stoppers are FDA compliant and biologically safe per USP Class VI non-cytotoxicity testing parameters; a certificate of conformance to regulatory standards is included. Platinum-cured silicone withstands temperatures from –60 to 460°F (–51 to 238°C). Autoclavable.