Cole-Parmer VapLock™ Solvent Delivery Cap with EPDM Air Inlet Valve, Two 1/4-28 ports, GL38; 1/EA
Representative image only.
Cole-Parmer Item # RK-12018-19
Reduce chemical evaporation and exposure to hazardous vapors
  • Use with semi-rigid tubing equal to or less than 1/8" (3.2 mm) OD
  • GL38 thread with two 1/4-28 ports
  • EPDM air inlet valve
PRODUCT OPTIONS
$57.31CAD / Each
SPECS & DESCRIPTION
Specifications & Description
  • Cap MaterialEPDM air inlet valve
  • Cap / Stopper SizeGL38
  • Cap ColorBlack/white
  • AutoclavableYes
  • Special Features2 x 1/4\"-28 UNF(F)
  • DescriptionSolvent Delivery Cap with EPDM Air Inlet Valve, two 1/4\"-28 ports, GL38; 1/ea
Key Features
  • Use with semi-rigid tubing equal to or less than 1/8" (3.2 mm) OD
  • GL38 thread with two 1/4-28 ports
  • EPDM air inlet valve
More About this Item
Create a closed system using solvent reservoir caps to prevent hazardous vapors from escaping into the lab. Caps accept standard 1/4"-28 UNF(M) fittings to easily connect semi-rigid tubing. Anti-twist design allows the inner body of the cap to rotate independently preventing tubes from tangling when caps are removed or attached. Caps are constructed of PTFE body and PP collar* and are autoclavable.
Caps with integrated air inlet valve and filter permit air to enter the bottle for vacuum displacement while solvent is pumped to the instrument. Valves are made of PTFE and 300-series SS components and feature a 50 µm PTFE filter to reduce particulate contamination. Select from FFKM, EPDM, or Viton® valve seal options.
*Does not come in contact with liquid under normal circumstances.