Spex VapLock™ Filter for VOC Vapors Inside Safety Cabinet; 2/pk
Representative image only.
Spex Item # RK-12021-10
Reduce harmful VOC vapors inside your safety cabinet
  • Adsorb VOC from cabinets storing flammable liquids
  • Flexible placement anywhere in cabinet with high strength magnets
  • Date notation area on label serves as a reminder of replacement
$155.81CAD / Pkg of 2
SPECS & DESCRIPTION
Specifications & Description
  • Width (in)2 1/4
  • Height (in)8 3/4
  • Housing MaterialStainless steel
  • SterileNo
  • ConnectionsMagnet
  • DescriptionFilter for VOC Vapors Inside Safety Cabinet; 2/pk
Key Features
  • Adsorb VOC from cabinets storing flammable liquids
  • Flexible placement anywhere in cabinet with high strength magnets
  • Date notation area on label serves as a reminder of replacement
More About this Item
The exhaust filter reduces harmful flammable VOC vapors found inside of safety cabinets. The filter is constructed of activated carbon medium and is contained in a stainless steel mesh cartridge. The filter features a strong 4” x 5.5” (10.2 x 13.9 cm) magnetic base to place anywhere within the cabinet. A date notation label is located on the bottom of the magnet as a reminder for replacement. The lifecycle of the filter varies upon your application.
Call Us At800-363-5900
Live chat
Bioz Article and Whitepaper Citations
Powered by Bioz See more details on Bioz