Cole-Parmer VapLock™ Breakthrough Indicator for Exhaust Filters; 1/ea
Representative image only.
Cole-Parmer Item # RK-12019-82
Use with exhaust filters to detect end-of-service life
  • Compatible with Cole-Parmer® VapLock™ activated carbon exhaust filters
PRODUCT OPTIONS
$76.17CAD / Each
SPECS & DESCRIPTION
Specifications & Description
  • MaterialAcrylic
  • Compatible WithVapLock™ activated carbon exhaust filters
  • DescriptionBreakthrough Indicator for Exhaust Filters; 1/ea
Key Features
  • Compatible with Cole-Parmer® VapLock™ activated carbon exhaust filters
More About this Item
Color change occurs when the indicator is exposed to organic solvent vapors. Indicator housing is made of acrylic.