This item is now discontinued. View the direct replacement: RK-61043-08.
Spex SamplePrep 6881 Grinding Mid-Size Vial set, 25 mL
Representative image only.
Spex SamplePrep Item # RK-04500-47
  • Everything you need to make one complete grinding vial
  • Sample capacity of 3 to 25 mL (1 to 40 g)
  • Easy to clean and reusable
  • Transparent center to view progress
SPECS & DESCRIPTION
Specifications & Description
  • Volume (mL)25
  • Tube Size25 mL
  • DescriptionGrinding Mid-Size Vial set, 25 mL
Includes
Stainless steel end plugs, stainless steel impactor, and polycarbonate center cylinder.
Key Features
  • Everything you need to make one complete grinding vial
  • Sample capacity of 3 to 25 mL (1 to 40 g)
  • Easy to clean and reusable
  • Transparent center to view progress
More About this Item
This mid-size grinding vial is perfect for samples such as tissue, seeds, polymers, pharmaceuticals, and textiles. The closed grinding vial prevents sample cross-contamination, retains volatiles, and protects hazardous and critical samples. The transparent polycarbonate center cylinder allows you to view the grinding process. Stainless steel end plugs and impactors are autoclavable and reusable; polycarbonate cylinders are disposable or may be cleaned wit hot water and detergent.