This item is now discontinued. Please browse similar items
Specwell M820-C Microscope/Telescopes with Built-In Reticle; 35x/8x, 7mm/6.5°
Representative image only.
Specwell Mfr # M820-C – Item # RK-03098-00
Versatile—magnify up to 35x or
view objects at a distance
  • Ideal for macroscopic quality control and inspection procedures
  • Includes a cross-hair reticle
  • 1 year warranty
SPECS & DESCRIPTION
Specifications & Description
  • Magnification8x/35x
  • Maximum Magnification25x
  • Head ConfigurationMonocular
  • Working Distance64 mm
  • Field of View7.0 mm
  • Height (InchShort)7.5156
  • Height (cm)19.1
  • Outside Diameter (in)1.25
  • Outside Diameter (cm)3.2
  • DescriptionMicroscope/telescopes with built-in reticle; magnification, 35x/8x; field of view
  • Warranty1 Year
1 Year Warranty
Includes
Neck strap, a micro lens stand, hard carrying case, soft vinly case, and carrying strap.
Key Features
  • Ideal for macroscopic quality control and inspection procedures
  • Includes a cross-hair reticle
  • 1 year warranty
More About this Item
Scopes are constructed of durable aluminum and plastic. Lenses come with covers to protect against scratches. Eyepiece has a soft edge for comfort and safety. Ideal for macroscopic quality control and inspection procedures.
This model features an adjustable focus distance from one foot at 17.5x to infinity at 8x.