This item is now discontinued. Please browse similar items

Spectra Por S/P 5 5 Standard RC Dialysis Tubing, 12-14 kDalton, 75 mm, 50ft/bx

Representative image only.
Spectra Por Mfr # 132754 – Item # RK-02900-54
Available in a variety of diameters and flat widths to meet your application needs
  • Regenerated cellulose (RC) membrane for the gentle separation and purification of solutes
  • 1 year warranty

SPECS & DESCRIPTION

Specifications & Description

  • MaterialRegenerated Cellulose
  • Max Molecular Weight Cut Off (Mwco) (Dalton)14000
  • Min Molecular Weight Cut Off (Mwco) (Dalton)12000
  • Flat Width (mm)75
  • Diameter (mm)48
  • Length Per Pack (ft)50
  • Diameter (in)1.89
  • Description5 Standard RC Dialysis Tubing, 12-14 kDalton, 75 mm, 50ft/bx
  • Warranty1 year
1 Year Warranty

Key Features

  • Regenerated cellulose (RC) membrane for the gentle separation and purification of solutes
  • 1 year warranty

More About this Item

MWCO of 12,000 to 14,000 Dalton

REVIEWS

Q&A

CUSTOMER SUPPORT

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At800-363-5900
Live chat