This item is now discontinued. Please browse similar items
Spectra Por S/P 1 5 Standard RC Dialysis Flat Disc, 6-8 kDalton, 100 mm dia, 50/pk
Representative image only.
Spectra Por Mfr # 132474 – Item # RK-02890-74
Available in a variety of dimensions to meet your application needs
  • Regenerated cellulose (RC) membrane for the gentle separation and purification of solutes
  • 1 year warranty
SPECS & DESCRIPTION
Specifications & Description
  • MaterialRegenerated Cellulose
  • Max Molecular Weight Cut Off (Mwco) (Dalton)8000
  • Min Molecular Weight Cut Off (Mwco) (Dalton)6000
  • Disc Diameter (mm)100
  • Description5 Standard RC Dialysis Flat Disc, 6-8 kDalton, 100 mm dia, 50/pk
  • Warranty1 year
1 Year Warranty
Key Features
  • Regenerated cellulose (RC) membrane for the gentle separation and purification of solutes
  • 1 year warranty
More About this Item
MWCO of 6000 to 8000 Dalton