This item is now discontinued. Please browse similar items
Spectra Por 132764 Magnetic weighted tubing closures; max gripping width, 55mm; 2/box
Representative image only.
Spectra Por Mfr # 132764 – Item # RK-02899-66
The most convenient method for sealing dialysis tubing without the risk of leaking
  • Easy to open and reseal
SPECS & DESCRIPTION
Specifications & Description
  • Tubing Sealing Width (mm)55
  • ColorRed
  • DescriptionMagnetic weighted tubing closures; max gripping width, 55mm; 2/box
Key Features
  • Easy to open and reseal
More About this Item
Magnetic closures contain a fluorocarbon-coated magnetic bar, eliminating the need for conventional stir bars. This closure provides complete submersion and gentle rotation of the sample on a magnetic stirrer.