This item is now discontinued. Please browse similar items
Solar Light PMA2170 Humidity/Temperature/Dew Point Sensor 1" handle, 0.5" sensor
Representative image only.
Solar Light Mfr # PMA2170 – Item # RK-97650-58
  • 1 year warranty
SPECS & DESCRIPTION
Specifications & Description
  • Resolution0.1% RH
  • FunctionMeasures relative humidity, ambient temperature, dew point
  • Intensity Range0 to 100% RH
  • Energy Range0 to 100% RH
  • DescriptionHumidity/Temperature/Dew Point Sensor 1\" handle, 0.5\" sensor
  • Warranty1 year
1 Year WarrantyNIST Traceable Calibration
Key Features
  • 1 year warranty
More About this Item
Replacement sensors for your Solar Light radiometer.
Measures relative humidity, ambient temperature and dew point.
Weatherproof detector housing available.