SHOWA 878-09 Gloves, Heavy-Weight, 28 mL Thickness, Large

Representative image only.
SHOWA Mfr # 878-09 – Item # RK-86357-21
Perfect for chemical handling, spill control, and hazmat applications

$96.95CAD / Pair of 2

BUY MORE & SAVE

  • 1-9$96.95CAD / Pair of 2
  • 10+$87.26CAD / Pair of 2

SPECS & DESCRIPTION

Specifications & Description

  • MaterialButyl
  • Thickness (mil)30;28
  • SizeLarge
  • Length (in)14
  • Length (cm)35
  • ColorBlack
  • GripSmooth
  • CuffRolled
  • ComplianceASTM F739
  • SterileNo
  • DescriptionGloves, heavy-weight, 28 mL thickness, large

More About this Item

These gloves provide excellent chemical resistance to hazardous materials — gases, aldehydes, amines, and ketones — without being bulky or restrictive. They are unlined to ensure an excellent grip. Gloves are 14" long with rolled cuffs that fit all hazmat suits.
Compliance: ASTM F-739

REVIEWS

Q&A

CUSTOMER SUPPORT

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At800-363-5900
Live chat